Learn More
Cayman Chemical AlocholIc AcId 10mg
SDP
Catalog No. 502988036
Encompass_Preferred
A bile acid and isomer of cholic acid; typically produced in the fetal or larval stages of development but has been found in adult mammals in the context of cancer or during liver regeneration in rats; a precursor to petromyzonol; levels are elevated in the plasma of mdr2-/- mice in a model of bile duct inflammation and hepatocellular carcinogenesis
Catalog No. 50-298-8036 Supplier Cayman Chemical Supplier No. 3041510 MG
A bile acid and isomer of cholic acid; typically produced in the fetal or larval stages of development but has been found in adult mammals in the context of cancer or during liver regeneration in rats; a precursor to petromyzonol; levels are elevated in the plasma of mdr2-/- mice in a model of bile duct inflammation and hepatocellular carcinogenesis