Learn More
Cayman Chemical AlocryptopIn 50mg
SDP
Catalog No. 502910718
Encompass_Preferred
An isoquinoline alkaloid; active against S. aureus, P. aeruginosa, and E. coli (MICs = 250, 125, and 125 UG/ml, respectively), P. falciparum and E. histolytica (IC50s = 5.08 and 135 µM, respectively), and a panel of seven phytopathogenic fungi (IC50s = 15.2-42.4 UG/ml)
Catalog No. 50-291-0718 Supplier Cayman Chemical Supplier No. 3475150 MG
An isoquinoline alkaloid; active against S. aureus, P. aeruginosa, and E. coli (MICs = 250, 125, and 125 UG/ml, respectively), P. falciparum and E. histolytica (IC50s = 5.08 and 135 µM, respectively), and a panel of seven phytopathogenic fungi (IC50s = 15.2-42.4 UG/ml)