Learn More
Cayman Chemical Diacerein, Purity: ≥98%, CAS: 13739-02-1, 250 mg
SDP
Catalog No. 502952089
Encompass_Preferred
A prodrug form of rhein; inhibits RANKL- and IL-1β-induced differentiation of RAW 264.7 cells into osteoclasts at 20 µg/ml; reduces levels of ICE, IL-1β, and IL-18 in the superficial zone of human osteoarthritic cartilage explants at 20 µg/ml; decreases carrageenan-, dextran-, or zymosan-induced paw edema in rats at 100 mg/kg
Catalog No. 50-295-2089 Supplier Cayman Chemical Supplier No. 11710250 MG
A prodrug form of rhein; inhibits RANKL- and IL-1β-induced differentiation of RAW 264.7 cells into osteoclasts at 20 µg/ml; reduces levels of ICE, IL-1β, and IL-18 in the superficial zone of human osteoarthritic cartilage explants at 20 µg/ml; decreases carrageenan-, dextran-, or zymosan-induced paw edema in rats at 100 mg/kg