Learn More
Cayman Chemical Aloe-emodin, Purity: ≥95%, CAS: 481-72-1, 10 mg
SDP
Catalog No. 502951927
Encompass_Preferred
Functions as a potent laxative by inducing chloride secretion in colonic mucosa and releasing acetylcholine, which stimulates contraction of intestinal smooth muscle; also demonstrates anti-tumor activity (ED50s = 1-13 µM), inducing apoptosis in various cancer cells
Catalog No. 50-295-1927 Supplier Cayman Chemical Supplier No. 1167710 MG
Functions as a potent laxative by inducing chloride secretion in colonic mucosa and releasing acetylcholine, which stimulates contraction of intestinal smooth muscle; also demonstrates anti-tumor activity (ED50s = 1-13 µM), inducing apoptosis in various cancer cells