Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aloe-emodin-8-O-β-D-glucopyranoside | 33037-46-6 | 5 MG
SDP

Catalog No. 5000421487
Encompass_Preferred

Aloe-emodin-8-O-β-D-glucopyranoside is a chemical compound primarily used as a laboratory reagent. Isolated from *Saussurea lappa*, it acts as a moderate inhibitor of human protein tyrosine phosphatase 1B (hPTP1B) with an IC₅₀ of 26.6 μM. This compound is intended for research purposes only.

  • Moderate inhibitor of human protein tyrosine phosphatase 1B (hPTP1B)
  • Isolated from *Saussurea lappa*
  • Exhibits cytotoxicity against MDR1 Pgp under-expressing human HCT116 cells (IC₅₀ 75 μM)
  • Exhibits cytotoxicity against MDR1 Pgp overexpressing human HepG2 cells (IC₅₀ > 100 μM)
  • High purity of 99.67%
  • Solid appearance with light yellow to yellow color
  • Molecular formula C₂₁H₂₀O₁₀
  • Molecular weight of 432.38

Catalog No. 50-004-21487 Supplier Medchemexpress LLC Supplier No. HYN24515MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aloe-emodin-8-O-β-D-glucopyranoside is a chemical compound primarily used as a laboratory reagent. Isolated from *Saussurea lappa*, it acts as a moderate inhibitor of human protein tyrosine phosphatase 1B (hPTP1B) with an IC₅₀ of 26.6 μM. This compound is intended for research purposes only.

  • Moderate inhibitor of human protein tyrosine phosphatase 1B (hPTP1B)
  • Isolated from *Saussurea lappa*
  • Exhibits cytotoxicity against MDR1 Pgp under-expressing human HCT116 cells (IC₅₀ 75 μM)
  • Exhibits cytotoxicity against MDR1 Pgp overexpressing human HepG2 cells (IC₅₀ > 100 μM)
  • High purity of 99.67%
  • Solid appearance with light yellow to yellow color
  • Molecular formula C₂₁H₂₀O₁₀
  • Molecular weight of 432.38

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.