Learn More
Medchemexpress LLC Aloe-emodin-8-O-β-D-glucopyranoside | 33037-46-6 | MFCD30573950 | 99.7% | 432.38 g·mol⁻¹ | C21H20O10 | 10 MG
SDP
Catalog No. 5000289689
Encompass_Preferred
Aloe-emodin-8-O-β-D-glucopyranoside is an anthraquinone glycoside isolated from Saussrurea lappa and supplied as a research reagent. It acts as a moderate inhibitor of human protein tyrosine phosphatase 1B (hPTP1B) with a reported IC50 of 26.6 μM, and is provided with analytical data suitable for laboratory use.
  • Anthraquinone glycoside with molecular formula C21H20O10.
  • Moderate inhibitor of hPTP1B (IC50 = 26.6 μM).
  • Molecular weight approximately 432.38 g·mol⁻¹.
  • High reported purity (about 99.67%).
  • Isolated from Saussrurea lappa and intended for research use.
  • Available in small-mass research pack sizes suitable for assay work.
Catalog No. 50-002-89689 Supplier Medchemexpress LLC Supplier No. HYN245110MG
Aloe-emodin-8-O-β-D-glucopyranoside is an anthraquinone glycoside isolated from Saussrurea lappa and supplied as a research reagent. It acts as a moderate inhibitor of human protein tyrosine phosphatase 1B (hPTP1B) with a reported IC50 of 26.6 μM, and is provided with analytical data suitable for laboratory use.
  • Anthraquinone glycoside with molecular formula C21H20O10.
  • Moderate inhibitor of hPTP1B (IC50 = 26.6 μM).
  • Molecular weight approximately 432.38 g·mol⁻¹.
  • High reported purity (about 99.67%).
  • Isolated from Saussrurea lappa and intended for research use.
  • Available in small-mass research pack sizes suitable for assay work.