Learn More
AdipoGen Aloe-Emodine (50 mg)
SDP
Supplier:  AdipoGen CDXA0658M050
Encompass_Preferred
Chemical. CAS: 481-72-1. Formula: C15H10O5. MW: 270.24. Aloe-emodin is a naturally anthraquinone derivative and an active ingredient of Aloe vera leaves and other plants. Aloe-emodin has been shown to exhibit many pharmacological effects, including anticancer, antivirus, anti-inflammatory, antibacterial, antiparasitic, neuroprotective, and hepatoprotective activities. It has potent laxative action by activating CFTR Cl- channels and inducing chloride secretion in colonic mucosa and releasing acetylcholine, which stimulates contraction of intestinal smooth muscle. It also demonstrates anti-tumor activity, inducing apoptosis in various cancer cells by increasing the production of reactive oxygen species (ROS) and induces apoptosis in glioma models through disruption of the mitochondria membrane potential.
Catalog No. 50-205-4291