Learn More
Description
Aloe Vera freeze dried powder is off-white to light beige in color, and has a fine homogenous texture. It has a light vegetable odor and is hygroscopic. Completely soluble in water (with a gel-like swelling) and insoluble in oil/solvents. Concentration of 200:1 compared to the fresh plant; to make a 1:1 solution suitable for an organic formula add 0.5g to 99.5g of purified water.
Alternative name(s): Aloe barbadensis
Specifications
Specifications
| Chemical Name or Material | Aloe Vera (Powder 200:1)(Freeze Dried) |
| Packaging | Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck |
| CAS | 85507-69-3 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | Not applicable |
| Quantity | 100 g |
| Grade | Ungraded |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
