Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Aloe Vera (Powder 200:1)(Freeze Dried)
SDP

Catalog No. 14186157 Shop All Medisca Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-157 500 g
14-185-964 100 g
14-185-587 5 g
14-185-529 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-186-157 Supplier Medisca Supplier No. 097708
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aloe Vera freeze dried powder is off-white to light beige in color, and has a fine homogenous texture. It has a light vegetable odor and is hygroscopic.

Aloe Vera freeze dried powder is off-white to light beige in color, and has a fine homogenous texture. It has a light vegetable odor and is hygroscopic. Completely soluble in water (with a gel-like swelling) and insoluble in oil/solvents. Concentration of 200:1 compared to the fresh plant; to make a 1:1 solution suitable for an organic formula add 0.5g to 99.5g of purified water.

Alternative name(s): Aloe barbadensis

Specifications

Chemical Name or Material Aloe Vera (Powder 200:1)(Freeze Dried)
Packaging Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 85507-69-3
Type Drug Standard
Molecular Weight (g/mol) Not applicable
Quantity 500 g
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.