Learn More
AdipoGen Aloeresin A (500 µg)
SDP
Supplier:  AdipoGen AGCN20439C500
Encompass_Preferred
Chemical. CAS: 74545-79-2. Formula: C28H28O11. MW: 540.5. Isolated from Aloe vera. Metabolite of aloesin. alpha-Glucosidase inhibitor. Antioxidant. Free radical scavenger.
Catalog No. 50-205-3607