Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aloeresin D | 105317-67-7 | MFCD32004290 | 99.5% | 556.56 | C29H32O11 | 10 MG
SDP

Catalog No. 5000296154
Encompass_Preferred

Aloeresin D is a chromone glycoside isolated from Aloe vera that inhibits β-secretase (BACE1) with a reported IC50 of 39 μM. It is supplied as a purified research compound with defined solubility and storage recommendations for biochemical and pharmacological studies.

  • Chromone glycoside isolated from Aloe vera.
  • Inhibits β-secretase (BACE1); IC50 = 39 μM.
  • High purity (≈99.5%).
  • Molecular formula C29H32O11; molecular weight 556.56 g/mol.
  • Soluble in DMSO (250 mg/mL); reported in vivo solubility ≥2.08 mg/mL.
  • Solid storage: 4°C, sealed, protected from moisture and light.

Catalog No. 50-002-96154 Supplier Medchemexpress LLC Supplier No. HYN221510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aloeresin D is a chromone glycoside isolated from Aloe vera that inhibits β-secretase (BACE1) with a reported IC50 of 39 μM. It is supplied as a purified research compound with defined solubility and storage recommendations for biochemical and pharmacological studies.

  • Chromone glycoside isolated from Aloe vera.
  • Inhibits β-secretase (BACE1); IC50 = 39 μM.
  • High purity (≈99.5%).
  • Molecular formula C29H32O11; molecular weight 556.56 g/mol.
  • Soluble in DMSO (250 mg/mL); reported in vivo solubility ≥2.08 mg/mL.
  • Solid storage: 4°C, sealed, protected from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.