Learn More
Enzo Life Sciences Aloesin (1mg). CAS: 30861-27-9
SDP
Supplier:  Enzo Life Sciences ALX350158M001
Encompass_Preferred
C-glycosylated chromone compound. Competitive inhibitor of tyrosinase. Shows weak free radical scavenging activity. Anti-inflammatory. Modulates melanogenesis (pigmentation-altering agent). Used in the cosmetic industry. Alternative name: Aloeresin B, NSC 631262, 8-beta-D-Glucopyranosyl-7-hydroxy-5-methyl-2-(2-oxopropyl)-4H-1-benzopyran-4-one. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8766