Learn More
Medchemexpress LLC HY-N2460 5mg Medchemexpress, Aloesin CAS:30861-27-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN24605MG
Encompass_Preferred
Medchemexpress, HY-N2460 5mg Aloesin CAS:30861-27-9 Aloesin (Aloeresin) is an active constituent of the herb aloe vera and displays anti-inflammatory activity, ultraviolet protection, and antibacterium effects. Aloesin exerts its anticancer effect through the MAPK signaling pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0396