Learn More
Selleck Chemical LLC Alogliptin (SYR-322) benzoate S2868-5mg
SDP
Catalog No. 503112333
Encompass_Preferred
Alogliptin (SYR-322) benzoate is a potent selective inhibitor of DPP-4 with IC50 of 10 nM exhibits greater than 10 000-fold selectivity over DPP-8 and DPP-9
Catalog No. 50-311-2333 Supplier Selleck Chemical LLC Supplier No. S28685MG
Alogliptin (SYR-322) benzoate is a potent selective inhibitor of DPP-4 with IC50 of 10 nM exhibits greater than 10 000-fold selectivity over DPP-8 and DPP-9