Learn More
TARGETMOL CHEMICALS INC ALOGLIPTIN BENZOATE 100MG
SDP
Catalog No. 502577131
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Alogliptin Benzoate (SYR 322)(SYR 322) an effective and specific DPP-4 inhibitor (IC50<10 nM) exhibits greater than 10 000-fold selectivity over DPP-8/9. Alogliptin may inhibit inflammatory responses by preventing the toll-like receptor 4 (TLR-4)-mediated formation of proinflammatory cytokines. purity: 99%
Catalog No. 50-257-7131 Supplier TARGETMOL CHEMICALS INC Supplier No. T2401100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Alogliptin Benzoate (SYR 322)(SYR 322) an effective and specific DPP-4 inhibitor (IC50<10 nM) exhibits greater than 10 000-fold selectivity over DPP-8/9. Alogliptin may inhibit inflammatory responses by preventing the toll-like receptor 4 (TLR-4)-mediated formation of proinflammatory cytokines. purity: 99%