Learn More
Medchemexpress LLC HY-A0023 5mg Medchemexpress, Alogliptin (Benzoate) CAS:850649-62-6 Purity:>98%
SDP
Catalog No. 502082307
Encompass_Preferred
Medchemexpress, HY-A0023 5mg Alogliptin (Benzoate) CAS:850649-62-6 Alogliptin Benzoate (SYR-322) is a potent, selective and orally active inhibitor of DPP-4 with an IC50 of <10 nM, and exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9. Alogliptin Benzoate can be used for the research of type 2 diabetes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2307 Supplier Medchemexpress LLC Supplier No. HYA00235MG
Medchemexpress, HY-A0023 5mg Alogliptin (Benzoate) CAS:850649-62-6 Alogliptin Benzoate (SYR-322) is a potent, selective and orally active inhibitor of DPP-4 with an IC50 of <10 nM, and exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9. Alogliptin Benzoate can be used for the research of type 2 diabetes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.