Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alogliptin benzoate | 850649-62-6 | 100.0% | 461.51 g·mol⁻¹ | C25H27N5O4 | 10 MG
SDP

Catalog No. 5000298444
Encompass_Preferred

Alogliptin benzoate is the benzoate salt of alogliptin, a potent and selective dipeptidyl peptidase-4 (DPP-4) inhibitor used for biochemical and preclinical research related to metabolic and cardiovascular disease. Supplied as a high-purity research reagent, it is intended for laboratory use only and not for human or clinical use.

  • Potent DPP-4 inhibitor with reported IC50 <10 nM.
  • High purity (HPLC) - 99.96%.
  • Molecular formula C25H27N5O4; molecular weight 461.51 g·mol⁻¹.
  • Available in small package sizes suitable for analytical work.
  • Store at 4°C sealed and away from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not intended for human use.

Catalog No. 50-002-98444 Supplier Medchemexpress LLC Supplier No. HYA002310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alogliptin benzoate is the benzoate salt of alogliptin, a potent and selective dipeptidyl peptidase-4 (DPP-4) inhibitor used for biochemical and preclinical research related to metabolic and cardiovascular disease. Supplied as a high-purity research reagent, it is intended for laboratory use only and not for human or clinical use.

  • Potent DPP-4 inhibitor with reported IC50 <10 nM.
  • High purity (HPLC) - 99.96%.
  • Molecular formula C25H27N5O4; molecular weight 461.51 g·mol⁻¹.
  • Available in small package sizes suitable for analytical work.
  • Store at 4°C sealed and away from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not intended for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.