Learn More
Medchemexpress LLC Alogliptin Benzoate | 850649-62-6 | 100.0% | C25H27N5O4 | 50 MG
SDP
Catalog No. 5000433501
Encompass_Preferred
Alogliptin Benzoate (SYR-322) is a potent, selective, and orally active inhibitor of DPP-4 with an IC50 of <10 nM, exhibiting greater than 10,000-fold selectivity over DPP-8 and DPP-9. It is used for the research of type 2 diabetes.
  • Potent and selective DPP-4 inhibitor
  • Exhibits over 10,000-fold selectivity over DPP-8 and DPP-9
  • Can be used for the research of type 2 diabetes
  • Inhibits LPS-induced extracellular signal-regulated kinase (ERK) phosphorylation
  • Reduces infarction area and improves brain vascular integrity in MCAO mice
Catalog No. 50-004-33501 Supplier Medchemexpress LLC Supplier No. HYA002350MG
Alogliptin Benzoate (SYR-322) is a potent, selective, and orally active inhibitor of DPP-4 with an IC50 of <10 nM, exhibiting greater than 10,000-fold selectivity over DPP-8 and DPP-9. It is used for the research of type 2 diabetes.
  • Potent and selective DPP-4 inhibitor
  • Exhibits over 10,000-fold selectivity over DPP-8 and DPP-9
  • Can be used for the research of type 2 diabetes
  • Inhibits LPS-induced extracellular signal-regulated kinase (ERK) phosphorylation
  • Reduces infarction area and improves brain vascular integrity in MCAO mice