Learn More
Apexbio Technology LLC Alogliptin Benzoate 850649-62-6 100mg
SDP
Catalog No. 5000093008
Encompass_Preferred
Alogliptin Benzoate (CAS 850649-62-6) is a potent and highly selective inhibitor of dipeptidyl peptidase-4 (DPP-4) with an IC50 below 10 nM By suppressing DPP-4 activity alogliptin elevates active GLP-1 levels leading to improved glycemic parameters In ob/ob mouse models chronic administration results in dose-dependent reductions in plasma DPP-4 activity non-fasting blood glucose glycated hemoglobin and triglycerides along with increased insulin secretion and pancreatic insulin content Alogliptin Benzoate is widely used in research focused on incretin biology glucose homeostasis and the pharmacological modulation of DPP-4
Catalog No. 50-000-93008 Supplier Apexbio Technology LLC Supplier No. A3156100
Alogliptin Benzoate (CAS 850649-62-6) is a potent and highly selective inhibitor of dipeptidyl peptidase-4 (DPP-4) with an IC50 below 10 nM By suppressing DPP-4 activity alogliptin elevates active GLP-1 levels leading to improved glycemic parameters In ob/ob mouse models chronic administration results in dose-dependent reductions in plasma DPP-4 activity non-fasting blood glucose glycated hemoglobin and triglycerides along with increased insulin secretion and pancreatic insulin content Alogliptin Benzoate is widely used in research focused on incretin biology glucose homeostasis and the pharmacological modulation of DPP-4