Learn More
Medchemexpress LLC Alogliptin (Benzoate) | 850649-62-6 | 99.7% | C25H27N5O4 | 25 MG
SDP
Catalog No. 5000359226
Encompass_Preferred
Alogliptin (Benzoate) is the analytical standard of Alogliptin (Benzoate), intended for research and analytical applications. It is a potent, selective, and orally active inhibitor of DPP-4 with an IC50 of <10 nM, exhibiting high selectivity over DPP-8 and DPP-9. This compound can be used for the research of type 2 diabetes and is suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Used for research and analytical applications
  • Potent, selective, and orally active inhibitor of DPP-4
  • Exhibits high selectivity over DPP-8 and DPP-9
  • Suitable for type 2 diabetes research
  • Ideal for HPLC, GC, and MS research experiments
Catalog No. 50-003-59226 Supplier Medchemexpress LLC Supplier No. 25MGHYA0023R
Alogliptin (Benzoate) is the analytical standard of Alogliptin (Benzoate), intended for research and analytical applications. It is a potent, selective, and orally active inhibitor of DPP-4 with an IC50 of <10 nM, exhibiting high selectivity over DPP-8 and DPP-9. This compound can be used for the research of type 2 diabetes and is suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Used for research and analytical applications
  • Potent, selective, and orally active inhibitor of DPP-4
  • Exhibits high selectivity over DPP-8 and DPP-9
  • Suitable for type 2 diabetes research
  • Ideal for HPLC, GC, and MS research experiments