Learn More
Medchemexpress LLC Alogliptin (Benzoate) | 850649-62-6 | 99.98% | C25H27N5O4 | 1 ML
SDP
Catalog No. 5000433160
Encompass_Preferred
Alogliptin Benzoate (SYR-322) is a potent, selective, and orally active inhibitor of DPP-4, primarily used for research related to type 2 diabetes. It exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9.
  • Potent and selective DPP-4 inhibitor
  • Orally active
  • Used for type 2 diabetes research
  • Inhibits LPS-induced ERK phosphorylation in U937 cells
  • Reduces infarction area and improves brain vascular integrity in MCAO mice
Catalog No. 50-004-33160 Supplier Medchemexpress LLC Supplier No. HYA002310MM/1ML
Alogliptin Benzoate (SYR-322) is a potent, selective, and orally active inhibitor of DPP-4, primarily used for research related to type 2 diabetes. It exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9.
  • Potent and selective DPP-4 inhibitor
  • Orally active
  • Used for type 2 diabetes research
  • Inhibits LPS-induced ERK phosphorylation in U937 cells
  • Reduces infarction area and improves brain vascular integrity in MCAO mice