Learn More
Medchemexpress LLC Alogliptin (benzoate) | 850649-62-6 | 99.98% | C25H27N5O4 | 500 MG
SDP
Catalog No. 5000433137
Encompass_Preferred
Alogliptin Benzoate (SYR-322) is a potent, selective, and orally active inhibitor of DPP-4 with an IC50 of <10 nM. It exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9 and can be used for the research of type 2 diabetes.
  • Potent, selective, and orally active DPP-4 inhibitor
  • High selectivity (greater than 10,000-fold) over DPP-8 and DPP-9
  • Useful for research into type 2 diabetes
Catalog No. 50-004-33137 Supplier Medchemexpress LLC Supplier No. HYA0023500MG
Alogliptin Benzoate (SYR-322) is a potent, selective, and orally active inhibitor of DPP-4 with an IC50 of <10 nM. It exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9 and can be used for the research of type 2 diabetes.
  • Potent, selective, and orally active DPP-4 inhibitor
  • High selectivity (greater than 10,000-fold) over DPP-8 and DPP-9
  • Useful for research into type 2 diabetes