Learn More
Medchemexpress LLC Alogliptin (Benzoate) | 850649-62-6 | C25H27N5O4 | 100 MG
SDP
Catalog No. 5000433503
Encompass_Preferred
Alogliptin Benzoate (SYR-322) is an analytical standard and a potent, selective, and orally active inhibitor of DPP-4. It exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9, making it suitable for type 2 diabetes research.
  • Potent and selective DPP-4 inhibitor
  • Orally active
  • Exhibits high selectivity over DPP-8 and DPP-9
  • Used for research of type 2 diabetes
  • Intended for research and analytical applications
Catalog No. 50-004-33503 Supplier Medchemexpress LLC Supplier No. HYA0023R100MG
Alogliptin Benzoate (SYR-322) is an analytical standard and a potent, selective, and orally active inhibitor of DPP-4. It exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9, making it suitable for type 2 diabetes research.
  • Potent and selective DPP-4 inhibitor
  • Orally active
  • Exhibits high selectivity over DPP-8 and DPP-9
  • Used for research of type 2 diabetes
  • Intended for research and analytical applications