Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alogliptin benzoate | 850649-62-6 | MFCD09833195 | 99.3% | 461.51 | C25H27N5O4 | 10 MG
SDP

Catalog No. 5000297910
Encompass_Preferred

Alogliptin benzoate (analytical standard) is the benzoate salt of the DPP-4 inhibitor alogliptin, provided as a high-purity reference material for analytical testing, method development, and assay validation. Supplied as a small solid quantity, it is suitable for chromatographic and mass spectrometry workflows.

  • High purity, typically 99.3%.
  • Provided as a solid in small, accurately measured quantities.
  • Suitable for HPLC and LC-MS method development and validation.
  • White to off-white crystalline powder, easy to handle.
  • Characterized by CAS 850649-62-6 and molecular formula C25H27N5O4.

Catalog No. 50-002-97910 Supplier Medchemexpress LLC Supplier No. HYA0023R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alogliptin benzoate (analytical standard) is the benzoate salt of the DPP-4 inhibitor alogliptin, provided as a high-purity reference material for analytical testing, method development, and assay validation. Supplied as a small solid quantity, it is suitable for chromatographic and mass spectrometry workflows.

  • High purity, typically 99.3%.
  • Provided as a solid in small, accurately measured quantities.
  • Suitable for HPLC and LC-MS method development and validation.
  • White to off-white crystalline powder, easy to handle.
  • Characterized by CAS 850649-62-6 and molecular formula C25H27N5O4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.