Learn More
Medchemexpress LLC Alogliptin benzoate (standard) | 850649-62-6 | 99.31% | C25H27N5O4 | 50 MG
SDP
Catalog No. 5000433139
Encompass_Preferred
Alogliptin benzoate (Standard) is the analytical standard of Alogliptin benzoate, intended for research and analytical applications. It is a potent, selective, and orally active inhibitor of DPP-4 with an IC50 of <10 nM, exhibiting high selectivity over DPP-8 and DPP-9. This compound is suitable for the research of type 2 diabetes.
  • Analytical standard for Alogliptin benzoate
  • Potent, selective, and orally active inhibitor of DPP-4
  • High selectivity over DPP-8 and DPP-9
  • Used for research of type 2 diabetes
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-33139 Supplier Medchemexpress LLC Supplier No. HYA0023R50MG
Alogliptin benzoate (Standard) is the analytical standard of Alogliptin benzoate, intended for research and analytical applications. It is a potent, selective, and orally active inhibitor of DPP-4 with an IC50 of <10 nM, exhibiting high selectivity over DPP-8 and DPP-9. This compound is suitable for the research of type 2 diabetes.
  • Analytical standard for Alogliptin benzoate
  • Potent, selective, and orally active inhibitor of DPP-4
  • High selectivity over DPP-8 and DPP-9
  • Used for research of type 2 diabetes
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS