Learn More
Medchemexpress LLC Alogliptin Benzoate (SYR 322) | 850649-62-6 | 100.0% | C25H27N5O4 | 200 MG
SDP
Catalog No. 5000432989
Encompass_Preferred
Alogliptin Benzoate (SYR-322) is a potent, selective, and orally active inhibitor of DPP-4 with an IC50 of <10 nM, exhibiting over 10,000-fold selectivity over DPP-8 and DPP-9. It is suitable for research into type 2 diabetes.
  • Potent, selective, and orally active inhibitor of DPP-4.
  • Inhibits LPS-induced extracellular signal-regulated kinase (ERK) phosphorylation in U937 cells.
  • Inhibits LPS-stimulated MMP-1 secretion and mRNA expression mediated by the ERK pathway in U937 cells.
  • Improves glucose tolerance and increases plasma insulin levels in female Wistar fatty rats.
  • Reduces infarction area and improves brain vascular integrity in middle cerebral artery occlusion (MCAO) mice.
Catalog No. 50-004-32989 Supplier Medchemexpress LLC Supplier No. HYA0023200MG
Alogliptin Benzoate (SYR-322) is a potent, selective, and orally active inhibitor of DPP-4 with an IC50 of <10 nM, exhibiting over 10,000-fold selectivity over DPP-8 and DPP-9. It is suitable for research into type 2 diabetes.
  • Potent, selective, and orally active inhibitor of DPP-4.
  • Inhibits LPS-induced extracellular signal-regulated kinase (ERK) phosphorylation in U937 cells.
  • Inhibits LPS-stimulated MMP-1 secretion and mRNA expression mediated by the ERK pathway in U937 cells.
  • Improves glucose tolerance and increases plasma insulin levels in female Wistar fatty rats.
  • Reduces infarction area and improves brain vascular integrity in middle cerebral artery occlusion (MCAO) mice.