Learn More
Medchemexpress LLC Alogliptin-d3 | 1133421-35-8 | 99.6% | C18H18D3N5O2 | 5 MG
SDP
Catalog No. 5000433226
Encompass_Preferred
Alogliptin-d3 is the deuterium labeled Alogliptin, a potent, selective, and orally active inhibitor of DPP-4 with an IC50 of <10 nM. It exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9 and can be used for the research of type 2 diabetes. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Deuterium labeled Alogliptin.
  • Potent, selective, and orally active inhibitor of DPP-4.
  • Exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9.
  • Suitable for research of type 2 diabetes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-33226 Supplier Medchemexpress LLC Supplier No. HYA0023AS15MG
Add to Cart
Add to Cart
Alogliptin-d3 is the deuterium labeled Alogliptin, a potent, selective, and orally active inhibitor of DPP-4 with an IC50 of <10 nM. It exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9 and can be used for the research of type 2 diabetes. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Deuterium labeled Alogliptin.
  • Potent, selective, and orally active inhibitor of DPP-4.
  • Exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9.
  • Suitable for research of type 2 diabetes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.