Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alogliptin-d3 (free base) | 1133421-35-8 | 99.6% | 342.41 g/mol | C18H18D3N5O2 | 10 MG
SDP

Catalog No. 5000298829
Encompass_Preferred

A deuterium-labeled analog of alogliptin supplied for research use as a tracer or internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS. It is intended for laboratory research and not for human use.

  • Deuterium-labeled tracer suitable for quantitative analysis.
  • High purity (99.6%).
  • Molecular weight 342.41 g/mol.
  • Supplied as a 10 mg solid, white to off-white.
  • Soluble in DMSO (100 mg/mL) and H2O (10 mg/mL) with ultrasonic or warming assistance.
  • Recommended storage: powder -20°C up to 3 years; in solvent -80°C up to 6 months.

Catalog No. 50-002-98829 Supplier Medchemexpress LLC Supplier No. HYA0023AS110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A deuterium-labeled analog of alogliptin supplied for research use as a tracer or internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS. It is intended for laboratory research and not for human use.

  • Deuterium-labeled tracer suitable for quantitative analysis.
  • High purity (99.6%).
  • Molecular weight 342.41 g/mol.
  • Supplied as a 10 mg solid, white to off-white.
  • Soluble in DMSO (100 mg/mL) and H2O (10 mg/mL) with ultrasonic or warming assistance.
  • Recommended storage: powder -20°C up to 3 years; in solvent -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.