Learn More
Medchemexpress LLC Alogliptin-13C,d3 benzoate | 1246817-18-4 | 98.8% | 1 MG
SDP
Catalog No. 5000432324
Encompass_Preferred
Alogliptin-13C,d3 benzoate is a stable isotope-labeled compound, primarily used as a tracer or an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. It is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Application as a tracer: Can be used in studies to track metabolic pathways or drug distribution.
  • Internal standard for quantitative analysis: Ideal for precise measurement in analytical methods such as NMR, GC-MS, or LC-MS due to its isotope labeling.
  • PROTAC linker: Serves as a linker in the construction of PROTACs, which are bifunctional molecules that induce targeted protein degradation.
  • Research use only: This product is intended for research and development purposes and not for sale to patients.
Catalog No. 50-004-32324 Supplier Medchemexpress LLC Supplier No. HYA0023AS21MG
Alogliptin-13C,d3 benzoate is a stable isotope-labeled compound, primarily used as a tracer or an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. It is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Application as a tracer: Can be used in studies to track metabolic pathways or drug distribution.
  • Internal standard for quantitative analysis: Ideal for precise measurement in analytical methods such as NMR, GC-MS, or LC-MS due to its isotope labeling.
  • PROTAC linker: Serves as a linker in the construction of PROTACs, which are bifunctional molecules that induce targeted protein degradation.
  • Research use only: This product is intended for research and development purposes and not for sale to patients.