Learn More
Cayman Chemical Alogliptin (benzoate salt), Purity: ≥98%, CAS: 850649-62-6, 100 mg
SDP
Catalog No. 502994946
Encompass_Preferred
An orally bioavailable DPP-4 inhibitor (IC50 = 6.9 nM) that is selective for DPP-4 over DPP-2, DPP-8, DPP-9, prolyl endopeptidase, FAP, and tryptase (IC50s = >100,000 nM); inhibits DPP-4 activity in vivo in rats, dogs, and cynomolgus monkeys (EC50s = 3.4, 4.9, and 5.6 ng/ml, respectively, in plasma); increases plasma GLP-1 and insulin levels and decreases blood glucose levels during an oral glucose challenge in Zucker fa/fa rats (10 mg/kg); decreases plasma DPP-4 activity and increases GLP-1 levels in diabetic ob/ob mice when administered for 29 days (2.8 mg/kg per day)
Catalog No. 50-299-4946 Supplier Cayman Chemical Supplier No. 23768100 MG
An orally bioavailable DPP-4 inhibitor (IC50 = 6.9 nM) that is selective for DPP-4 over DPP-2, DPP-8, DPP-9, prolyl endopeptidase, FAP, and tryptase (IC50s = >100,000 nM); inhibits DPP-4 activity in vivo in rats, dogs, and cynomolgus monkeys (EC50s = 3.4, 4.9, and 5.6 ng/ml, respectively, in plasma); increases plasma GLP-1 and insulin levels and decreases blood glucose levels during an oral glucose challenge in Zucker fa/fa rats (10 mg/kg); decreases plasma DPP-4 activity and increases GLP-1 levels in diabetic ob/ob mice when administered for 29 days (2.8 mg/kg per day)