Learn More
AdipoGen Aloin (100 mg)
SDP
Supplier:  AdipoGen CDXA0527M100
Encompass_Preferred
Chemical. CAS: 1415-73-2. Formula: C21H22O9. MW: 418.39. Isolated from plant source. Aloin is a naturally occurring substance found in various Aloe plant species. Aloin was originally used as a laxative in the treatment of constipation. Aloin is a chemopreventive and antineoplastic agent. It is a cytotoxic against cancer cells. As a anticancer agent it inhibits Topoisomerase II alpha, induces apoptosis, targets High Mobility Group Box 1 (HMGB1) and inhibits angiogenesis. It has been shown to be a potent skin depigmenting agent, enhancing melanogenesis. It has been described as a tyroniase inhibitor. It regulates transglutaminase activity. In addition Aloin is an anti-inflammatory agent, inhibiting the NF-kappaB pathway, antitrypanosomal, iron chelating and microbiota modulating compound.
Catalog No. 50-205-4275