Learn More
Medchemexpress LLC HY-N6013 1mg Medchemexpress, Aloin(mixture of A&B) CAS:8015-61-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN60131MG
Encompass_Preferred
Medchemexpress, HY-N6013 1mg Aloin(mixture of A&B) CAS:8015-61-0 Aloin (mixture of A&B) is anthraquinone derivative isolated from Aloe vera. Aloin (mixture of A&B) has diverse biological activities such as anti-inflammatory, immunity, antidiabetic, antioxidant, antibacterial, antifungal, and antitumor activities. Aloin (mixture of A&B) also an effective inhibitor of stimulated granulocyte matrix metalloproteinases (MMPs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2943