Learn More
Cayman Chemical AloIsIn RP106 10mg
SDP
Catalog No. 502986034
Encompass_Preferred
An inhibitor of Cdk1, Cdk5, and GSK3 (IC50s = 0.70, 1.5, and 0.92 µM, respectively); a derivative of the aloisines A and B
Catalog No. 50-298-6034 Supplier Cayman Chemical Supplier No. 2187310 MG
An inhibitor of Cdk1, Cdk5, and GSK3 (IC50s = 0.70, 1.5, and 0.92 µM, respectively); a derivative of the aloisines A and B