Learn More
Cayman Chemical Alloisolithocholic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2410277-60-8, 5 mg
SDP
Catalog No. 502944780
Encompass_Preferred
An internal standard for the quantification of alloisolithocholic acid by GC- or LC-MS
Catalog No. 50-294-4780 Supplier Cayman Chemical Supplier No. 411555 MG
An internal standard for the quantification of alloisolithocholic acid by GC- or LC-MS