Learn More
Cayman Chemical Allolithocholic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), 10 mg
SDP
Catalog No. 502944621
Encompass_Preferred
An internal standard for the quantification of allolithocholic acid by GC- or LC-MS
Catalog No. 50-294-4621 Supplier Cayman Chemical Supplier No. 4115610 MG
An internal standard for the quantification of allolithocholic acid by GC- or LC-MS