Learn More
Apexbio Technology LLC Aloperine 56293-29-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062274
Encompass_Preferred
Aloperine is a naturally derived alkaloid known for its biological properties including anti-inflammatory antiviral antibacterial and antitumor activities Mechanistic studies suggest that aloperine exerts anti-inflammatory effects through inhibition of inflammatory mediators and inflammatory signaling pathways such as nuclear factor-kappa B (NF- B) and mitogen-activated protein kinase (MAPK) pathways In vitro assays demonstrate antiviral potential related to inhibition of specific viral replication stages Additionally its antitumor properties have been identified in multiple cancer cell lines where aloperine can induce apoptosis inhibit proliferation and migration and modulate cell cycle progression Aloperine typically exhibits IC50 values ranging from low micromolar to sub-micromolar concentrations depending on cell lines and experimental settings It serves as a research tool compound in immunological oncological and infectious disease studies
Catalog No. 50-000-62274 Supplier Apexbio Technology LLC Supplier No. B16545.1
Aloperine is a naturally derived alkaloid known for its biological properties including anti-inflammatory antiviral antibacterial and antitumor activities Mechanistic studies suggest that aloperine exerts anti-inflammatory effects through inhibition of inflammatory mediators and inflammatory signaling pathways such as nuclear factor-kappa B (NF- B) and mitogen-activated protein kinase (MAPK) pathways In vitro assays demonstrate antiviral potential related to inhibition of specific viral replication stages Additionally its antitumor properties have been identified in multiple cancer cell lines where aloperine can induce apoptosis inhibit proliferation and migration and modulate cell cycle progression Aloperine typically exhibits IC50 values ranging from low micromolar to sub-micromolar concentrations depending on cell lines and experimental settings It serves as a research tool compound in immunological oncological and infectious disease studies