Learn More
Medchemexpress LLC HY-13516 50mg Medchemexpress, Aloperine CAS:56293-29-9 Purity:>98%
SDP
Catalog No. 501969865
Encompass_Preferred
Medchemexpress, HY-13516 50mg Aloperine CAS:56293-29-9 Aloperine is an alkaloid in sophora plants such as Sophora alopecuroides L, which has shown anti-cancer, anti-inflammatory and anti-virus properties. Aloperine is widely used to treat patients with allergic contact dermatitis eczema and other skin inflammation in China. Aloperine induces apoptosis and autophagy in HL-60 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9865 Supplier Medchemexpress LLC Supplier No. HY1351650MG
Medchemexpress, HY-13516 50mg Aloperine CAS:56293-29-9 Aloperine is an alkaloid in sophora plants such as Sophora alopecuroides L, which has shown anti-cancer, anti-inflammatory and anti-virus properties. Aloperine is widely used to treat patients with allergic contact dermatitis eczema and other skin inflammation in China. Aloperine induces apoptosis and autophagy in HL-60 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.