Learn More
Cayman Chemical Allopurinol Riboside, Purity: ≥95%, CAS: 16220-07-8, 10 mg
SDP
Catalog No. 502950877
Encompass_Preferred
A ribonucleoside; inhibits Leishmania promastigote growth and prevents amastigote maturation into promastigotes; prevents mortality in mouse models of T. cruzi infection at 239, 511, and 929 mg/kg per day
Catalog No. 50-295-0877 Supplier Cayman Chemical Supplier No. 3345910 MG
A ribonucleoside; inhibits Leishmania promastigote growth and prevents amastigote maturation into promastigotes; prevents mortality in mouse models of T. cruzi infection at 239, 511, and 929 mg/kg per day