Learn More
Medchemexpress LLC Alosetron-d3 hydrochloride | 1189919-71-8 | 99.0% | 333.83 g/mol | C17H16D3ClN4O | 10 MG
SDP
Catalog No. 5000298634
Encompass_Preferred
Alosetron-d3 hydrochloride is a deuterium-labeled analogue of alosetron supplied as the hydrochloride salt. It is intended for use as an internal standard or tracer in quantitative analytical methods, including LC-MS, GC-MS, and NMR, for research applications.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for LC-MS, GC-MS, and NMR methods.
  • High purity for analytical consistency.
  • Supplied as a hydrochloride salt for improved stability and solubility.
  • Provided in milligram quantities appropriate for analytical workflows.
Catalog No. 50-002-98634 Supplier Medchemexpress LLC Supplier No. HY70050CS10MG
Alosetron-d3 hydrochloride is a deuterium-labeled analogue of alosetron supplied as the hydrochloride salt. It is intended for use as an internal standard or tracer in quantitative analytical methods, including LC-MS, GC-MS, and NMR, for research applications.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for LC-MS, GC-MS, and NMR methods.
  • High purity for analytical consistency.
  • Supplied as a hydrochloride salt for improved stability and solubility.
  • Provided in milligram quantities appropriate for analytical workflows.