Learn More
Medchemexpress LLC Alosetron-d3 hydrochloride | 1189919-71-8 | 99.0% | C17H16D3ClN4O | 5 MG
SDP
Catalog No. 5000434810
Encompass_Preferred
Alosetron-d3 hydrochloride is a deuterium labeled Alosetron, functioning as a serotonin 5HT3-receptor antagonist. It is suitable for use as a tracer and an internal standard in NMR, GC-MS, or LC-MS analysis for quantitative applications.
  • Deuterium labeled compound
  • Serotonin 5HT3-receptor antagonist
  • Internal standard for quantitative analysis
  • Used in neurological disease research
  • Supports drug concentration detection
Catalog No. 50-004-34810 Supplier Medchemexpress LLC Supplier No. HY70050CS5MG
Alosetron-d3 hydrochloride is a deuterium labeled Alosetron, functioning as a serotonin 5HT3-receptor antagonist. It is suitable for use as a tracer and an internal standard in NMR, GC-MS, or LC-MS analysis for quantitative applications.
  • Deuterium labeled compound
  • Serotonin 5HT3-receptor antagonist
  • Internal standard for quantitative analysis
  • Used in neurological disease research
  • Supports drug concentration detection