Learn More
Apexbio Technology LLC Alosetron Hydrochloride 122852-69-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000095170
Encompass_Preferred
Alosetron Hydrochloride (CAS 122852-69-1) is a potent antagonist of the serotonin 5-HT3 receptor a ligand-gated ion channel implicated in the modulation of gastrointestinal function By selectively inhibiting 5-HT3 receptor-mediated signaling on enteric neurons Alosetron modulates visceral sensation and gastrointestinal motility This mechanism underlies its application in the study of serotonergic pathways involved in irritable bowel syndrome and related gastrointestinal disorders Alosetron serves as a valuable tool in biomedical research investigating the neurogastroenterology of serotonergic signaling and its role in gut-brain axis modulation
Catalog No. 50-000-95170 Supplier Apexbio Technology LLC Supplier No. A31595.1
Alosetron Hydrochloride (CAS 122852-69-1) is a potent antagonist of the serotonin 5-HT3 receptor a ligand-gated ion channel implicated in the modulation of gastrointestinal function By selectively inhibiting 5-HT3 receptor-mediated signaling on enteric neurons Alosetron modulates visceral sensation and gastrointestinal motility This mechanism underlies its application in the study of serotonergic pathways involved in irritable bowel syndrome and related gastrointestinal disorders Alosetron serves as a valuable tool in biomedical research investigating the neurogastroenterology of serotonergic signaling and its role in gut-brain axis modulation