Learn More
Medchemexpress LLC Alosetron Hydrochloride | 122852-69-1 | 99.6% | C17H19ClN4O | 10 MM * 1 ML
SDP
Catalog No. 5000433954
Encompass_Preferred
Alosetron Hydrochloride (GR 68755C) is a potent and highly selective serotonin 5-HT3 receptor antagonist, utilized for irritable bowel syndrome (IBS) research. It blocks 5-HT3-mediated depolarisation of guinea-pig myenteric and submucosal neurons (IC50 ~55 nM), attenuates visceral nociceptive effects, and demonstrates anti-inflammatory effects.
  • Potent and highly selective serotonin 5-HT3 receptor antagonist.
  • Used for research of irritable bowel syndrome.
  • Blocks 5HT3-mediated depolarisation of neurons.
  • Attenuates visceral nociceptive effects.
  • Demonstrates anti-inflammatory effects.
Catalog No. 50-004-33954 Supplier Medchemexpress LLC Supplier No. HY70050C10MM/1ML
Alosetron Hydrochloride (GR 68755C) is a potent and highly selective serotonin 5-HT3 receptor antagonist, utilized for irritable bowel syndrome (IBS) research. It blocks 5-HT3-mediated depolarisation of guinea-pig myenteric and submucosal neurons (IC50 ~55 nM), attenuates visceral nociceptive effects, and demonstrates anti-inflammatory effects.
  • Potent and highly selective serotonin 5-HT3 receptor antagonist.
  • Used for research of irritable bowel syndrome.
  • Blocks 5HT3-mediated depolarisation of neurons.
  • Attenuates visceral nociceptive effects.
  • Demonstrates anti-inflammatory effects.