Learn More
Medchemexpress LLC Alosetron Hydrochloride | 122852-69-1 | 99.6% | C17H19ClN4O | 100 MG
SDP
Catalog No. 5000434584
Encompass_Preferred
Alosetron Hydrochloride is a potent and highly selective serotonin 5-HT3 receptor antagonist used in research for irritable bowel syndrome (IBS). It blocks the fast 5HT3-mediated depolarization of guinea-pig myenteric and submucosal neurons, with an IC50 of approximately 55 nM. It also attenuates visceral nociceptive effects of rectal distension in dogs and demonstrates anti-inflammatory properties.
  • Potent and highly selective serotonin 5-HT3 receptor antagonist
  • Applicable for research in irritable bowel syndrome (IBS)
  • Blocks fast 5HT3-mediated depolarization of guinea-pig myenteric and submucosal neurons, with an IC50 of approximately 55 nM
  • Attenuates visceral nociceptive effects of rectal distension in dogs
  • Demonstrates anti-inflammatory effects
Catalog No. 50-004-34584 Supplier Medchemexpress LLC Supplier No. HY70050C100MG
Alosetron Hydrochloride is a potent and highly selective serotonin 5-HT3 receptor antagonist used in research for irritable bowel syndrome (IBS). It blocks the fast 5HT3-mediated depolarization of guinea-pig myenteric and submucosal neurons, with an IC50 of approximately 55 nM. It also attenuates visceral nociceptive effects of rectal distension in dogs and demonstrates anti-inflammatory properties.
  • Potent and highly selective serotonin 5-HT3 receptor antagonist
  • Applicable for research in irritable bowel syndrome (IBS)
  • Blocks fast 5HT3-mediated depolarization of guinea-pig myenteric and submucosal neurons, with an IC50 of approximately 55 nM
  • Attenuates visceral nociceptive effects of rectal distension in dogs
  • Demonstrates anti-inflammatory effects