Learn More
Medchemexpress LLC Alosetron Hydrochloride | 122852-69-1 | 99.6% | C17H19ClN4O | 25 MG
SDP
Catalog No. 5000357634
Encompass_Preferred
Alosetron Hydrochloride is a potent and highly selective serotonin 5-HT3 receptor antagonist used for the research of irritable bowel syndrome (IBS). It blocks fast 5HT3-mediated depolarization of guinea-pig myenteric and submucosal neurons, and attenuates visceral nociceptive effects.
  • Potent and highly selective serotonin 5-HT3 receptor antagonist
  • Used for research of irritable bowel syndrome (IBS)
  • Blocks fast 5HT3-mediated depolarization of myenteric and submucosal neurons
  • Attenuates visceral nociceptive effect of rectal distension
  • Exhibits anti-inflammatory effects
Catalog No. 50-003-57634 Supplier Medchemexpress LLC Supplier No. 25MGHY70050C
Alosetron Hydrochloride is a potent and highly selective serotonin 5-HT3 receptor antagonist used for the research of irritable bowel syndrome (IBS). It blocks fast 5HT3-mediated depolarization of guinea-pig myenteric and submucosal neurons, and attenuates visceral nociceptive effects.
  • Potent and highly selective serotonin 5-HT3 receptor antagonist
  • Used for research of irritable bowel syndrome (IBS)
  • Blocks fast 5HT3-mediated depolarization of myenteric and submucosal neurons
  • Attenuates visceral nociceptive effect of rectal distension
  • Exhibits anti-inflammatory effects