Learn More
Medchemexpress LLC Alosetron Hydrochloride | 122852-69-1 | 99.63% | C17H19ClN4O | 50 MG
SDP
Catalog No. 5000434360
Encompass_Preferred
Alosetron Hydrochloride is a potent and highly selective antagonist of the serotonin 5-HT3 receptor. It is researched for its application in irritable bowel syndrome (IBS). The compound effectively blocks the rapid 5HT3-mediated depolarization in guinea-pig myenteric and submucosal neurons, with an IC50 of approximately 55 nM. It also demonstrates an ability to lessen the visceral nociceptive effects of rectal distension in conscious or anesthetized dogs, and exhibits anti-inflammatory properties.
  • Highly selective antagonist of the serotonin 5-HT3 receptor.
  • Under investigation for irritable bowel syndrome.
  • Blocks 5HT3-mediated depolarization.
  • Attenuates visceral nociceptive effects.
  • Exhibits anti-inflammatory effects.
Catalog No. 50-004-34360 Supplier Medchemexpress LLC Supplier No. HY70050C50MG
Alosetron Hydrochloride is a potent and highly selective antagonist of the serotonin 5-HT3 receptor. It is researched for its application in irritable bowel syndrome (IBS). The compound effectively blocks the rapid 5HT3-mediated depolarization in guinea-pig myenteric and submucosal neurons, with an IC50 of approximately 55 nM. It also demonstrates an ability to lessen the visceral nociceptive effects of rectal distension in conscious or anesthetized dogs, and exhibits anti-inflammatory properties.
  • Highly selective antagonist of the serotonin 5-HT3 receptor.
  • Under investigation for irritable bowel syndrome.
  • Blocks 5HT3-mediated depolarization.
  • Attenuates visceral nociceptive effects.
  • Exhibits anti-inflammatory effects.