Learn More
Medchemexpress LLC Alosetron hydrochloride | 122852-69-1 | MFCD03453647 | 99.6% | 10 MG
SDP
Catalog No. 5000298470
Encompass_Preferred
Alosetron hydrochloride is a potent, selective 5-HT3 receptor antagonist provided for research use in studies of irritable bowel syndrome and visceral nociception. It is supplied as a high-purity solid and is also available as ready-to-use 10 mM solutions for laboratory assays.
  • Potent and selective 5-HT3 receptor antagonist.
  • Used in research on diarrhea-predominant irritable bowel syndrome and visceral nociception.
  • High purity suitable for analytical and in vitro applications.
  • Available as a solid and as pre-made 10 mM solutions for convenience.
  • Characterization includes CAS 122852-69-1 and molecular weight 330.81 g/mol.
Catalog No. 50-002-98470 Supplier Medchemexpress LLC Supplier No. HY70050C10MG
Alosetron hydrochloride is a potent, selective 5-HT3 receptor antagonist provided for research use in studies of irritable bowel syndrome and visceral nociception. It is supplied as a high-purity solid and is also available as ready-to-use 10 mM solutions for laboratory assays.
  • Potent and selective 5-HT3 receptor antagonist.
  • Used in research on diarrhea-predominant irritable bowel syndrome and visceral nociception.
  • High purity suitable for analytical and in vitro applications.
  • Available as a solid and as pre-made 10 mM solutions for convenience.
  • Characterization includes CAS 122852-69-1 and molecular weight 330.81 g/mol.