Learn More
Selleck Chemical LLC Alosetron Hydrochloride S4694-20mg
SDP
Catalog No. 503108427
Encompass_Preferred
Alosetron Hydrochloride (GR 68755C GR 68755X Lotronex) is the hydrochloride salt form of alosetron a potent and selective 5-HT3 receptor antagonist
Catalog No. 50-310-8427 Supplier Selleck Chemical LLC Supplier No. S469420MG
Alosetron Hydrochloride (GR 68755C GR 68755X Lotronex) is the hydrochloride salt form of alosetron a potent and selective 5-HT3 receptor antagonist