Learn More
Cayman Chemical Alosetron (hydrochloride), Purity: ≥98%, CAS: 122852-69-1, 100 mg
SDP
Catalog No. 502987871
Encompass_Preferred
A 5-HT3 receptor antagonist (Kis = 0.3 and 0.4 nM for the rat cortical and human recombinant receptors, respectively); selective for 5-HT3 over 5-HT2 and 5-HT1A receptors (Kis = 6.3 and >100 µM, respectively), as well as α1-, β1-, and β2- adrenergic, M1, M2, and M3 muscarinic, GABAA, dopamine D2, and nicotinic receptors at 10 μM; inhibits 5-HT-induced depolarization of isolated rat vagus nerve at 0.3-10 nM; reduces wrap restraint stress-induced defecation and the number of abdominal muscle contractions in a rat model of IBS-D at 0.1 mg/kg
Catalog No. 50-298-7871 Supplier Cayman Chemical Supplier No. 22434100 MG
A 5-HT3 receptor antagonist (Kis = 0.3 and 0.4 nM for the rat cortical and human recombinant receptors, respectively); selective for 5-HT3 over 5-HT2 and 5-HT1A receptors (Kis = 6.3 and >100 µM, respectively), as well as α1-, β1-, and β2- adrenergic, M1, M2, and M3 muscarinic, GABAA, dopamine D2, and nicotinic receptors at 10 μM; inhibits 5-HT-induced depolarization of isolated rat vagus nerve at 0.3-10 nM; reduces wrap restraint stress-induced defecation and the number of abdominal muscle contractions in a rat model of IBS-D at 0.1 mg/kg