Learn More
Aviva Systems Biology Corp ALOX12 Antibody - C-terminal region (ARP54350_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54350_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALOX12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ALOX12. Alternate names: 12-LOX, LOG12, 12S-LOXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61183