Learn More
Aviva Systems Biology Corp ALOX15 Antibody - middle region (ARP56030_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56030_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALOX15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ALOX15. Alternate names: 15-LOX2, 15LOX-1, 15-LOX-1ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61700